Learn More
Tosoh Bioscience LLC TSKGEL GUARD FOR SW 8X40
Supplier:  Tosoh Bioscience LLC 0008805
Encompass
Column, Guard; For 8mm I.D. Tosoh TSKgel SW glass columns; Media: Silica; 8mm I.D. x 4cm Length
Catalog No. 50-851-289