Learn More
Tosoh Bioscience LLC TSKGEL GUARD G1000HXL GMHXL-L
Supplier:  Tosoh Bioscience LLC 0007113
Encompass
Column, Guard; For 6mm I.D. Tosoh TSKgel G1000Hxl-GMHxl-L; Media: Polymer; Particle: 5-6 micrometer; 6mm I.D. x 4cm Length
Catalog No. 50-851-159