Learn More
Tosoh Bioscience LLC TSKGEL GUARD G5000HXL-GMHXL
Supplier:  Tosoh Bioscience LLC 0013727
Encompass
Column, Guard; For 9 micrometer-13 micrometer Tosoh TSKgel G5000HXL-GMHXL type columns; Media: Polymer; 6mm I.D. x 4cm Length
Catalog No. 50-851-312