Learn More
Tosoh Bioscience LLC TSKGEL GUARD PWXL AND G-DNA-PW
Supplier:  Tosoh Bioscience LLC 0008033
Encompass
Column, Guard; For 7.8mm I.D. Tosoh TSKgel PWxl; Particle: 12 micrometer; Media: Polymer; 6mm I.D. x 4cm Length
Catalog No. 50-851-254