Learn More
Tosoh Bioscience LLC TSKGEL HICPAK PW (30) 2X25ML
Supplier:  Tosoh Bioscience LLC 0043175
Encompass
Media, Bulk Resins; LabPak Sampler for Hydrophobic Interaction Chromatography; Tosoh TSKgel HICPAK PW (Ether-5PW, Phenyl-5PW); Media: Polymer; Particle: 30 micrometer; 2 x 25mL
Catalog No. 50-851-724