Learn More
Tosoh Bioscience LLC TSKGEL IC-CATION SW 4.6X50
Supplier:  Tosoh Bioscience LLC 0008055
Encompass
Column, Ion exchange chromatography; Tosoh; TSKgel IC; Cation SW; Media: hydrophilic silica gel; Particle: 5u m; 4.6mm I.D. x 5cm length
Catalog No. 50-851-262