Learn More
Tosoh Bioscience LLC TSKGEL IEXPAK PW (20) 3X25ML
Supplier:  Tosoh Bioscience LLC 0043380
Encompass
Media, Bulk anion exchange resins; Tosoh; TSK-GEL; LabPak sampler for ion exchange chromatography; Long column life; IEXPAK PW (20) (DEAE-5PW, SP-5PW, SuperQ-5PW); Media: polymer; Particle: 20u m; 3 x 25mL
Catalog No. 50-851-743