Learn More
Tosoh Bioscience LLC TSKGEL NH2-100 2X50
Catalog No. 50550544 Shop All Tosoh Bioscience LLC Products
Encompass
LC Hydrophilic Columns, TSKgel NH2-100, 2 mm ID x 5 cm, 3 um
Catalog No. 50-550-544 Supplier Tosoh Bioscience LLC Supplier No. 0021967
LC Hydrophilic Columns, TSKgel NH2-100, 2 mm ID x 5 cm, 3 um