Learn More
Tosoh Bioscience LLC TSKGEL OAPAK-A 7.8X300
Supplier:  Tosoh Bioscience LLC 0016653
Encompass
Column, Ion exchange chromatography; Tosoh; TSKgel OApak-A; Matrix: hydrophilic polymer; Stainless-steel; Counter ion: Hsup+ Particle size: 5u m; pH: 2.0 -12; 7.8mm I.D. x 30cm length
Catalog No. 50-851-422