Learn More
Tosoh Bioscience LLC TSKGEL OCTADECYL-2PW 2X150
Supplier: Tosoh Bioscience LLC 0018754
Column, Reversed Phase Chromatography; Tosoh TSKgel Octadecyl-2PW; Media: Polymer; Particle: 5 micrometer; Pore Size: 100-200 angstrom; 2mm I.D. x 15cm Length; Stainless Steel