Learn More
Tosoh Bioscience LLC TSKGEL OCTADECYL-2PW 6X150
Catalog No. 50851455 Shop All Tosoh Bioscience LLC Products
Encompass
Column, Reversed Phase Chromatography; Tosoh TSKgel Octadecyl-2PW; Media: Polymer; Particle: 5 micrometer; Pore Size: 100-200 angstrom; 6mm I.D. x 15cm Length; Stainless Steel
Catalog No. 50-851-455 Supplier Tosoh Bioscience LLC Supplier No. 0017501
Column, Reversed Phase Chromatography; Tosoh TSKgel Octadecyl-2PW; Media: Polymer; Particle: 5 micrometer; Pore Size: 100-200 angstrom; 6mm I.D. x 15cm Length; Stainless Steel