Learn More
Tosoh Bioscience LLC TSKGEL OCTADECYL-4PW 2X10 3PK
Supplier:  Tosoh Bioscience LLC 0042160
Encompass
Cartridge, Guard; For 2mm I.D. Tosoh TSKgel Octadecyl-4PW column; Media: Polymer; 2mm I.D. x 1cm Length; 3/Pk
Catalog No. 50-851-708