Learn More
Tosoh Bioscience LLC TSKGEL OCTADECYL-4PW 4.6X150
Supplier:  Tosoh Bioscience LLC 0013351
Encompass
Column, Reversed Phase Chromatography; Tosoh TSKgel Octadecyl-4PW; Media: Polymer; Particle: 7 micrometer; Pore Size: 500 angstrom; 4.6mm I.D. x 15cm Length; Stainless Steel
Catalog No. 50-851-309