Learn More
Tosoh Bioscience LLC TSKGEL ODS-100V 2X10 3UM 1/PK
Supplier: Tosoh Bioscience LLC 0021814
Column, Reversed Phase Chromatography; Tosoh TSKgel ODS-100V; Media: Silica; Particle: 3 micrometer; Pore Size: 100 angstrom; 2.0mm I.D. x 1cm Length; Stainless Steel; 3/Pk