Learn More
Tosoh Bioscience LLC TSKGEL ODS-100V 2X750 3UM
Supplier:  Tosoh Bioscience LLC 0021811
Encompass
Column, Reversed Phase Chromatography; Tosoh TSKgel ODS-100V; Media: Silica; Particle: 3 micrometer; Pore Size: 100 angstrom;2.0mm I.D. x 7.5cm Length; Stainless Steel
Catalog No. 50-851-697