Learn More
Tosoh Bioscience LLC TSKGEL ODS-100V 4.6X150 5UM
Supplier:  Tosoh Bioscience LLC 0021455
Encompass
Column, Reversed Phase Chromatography; Tosoh TSKgel ODS-100V; Media: Silica; Particle: 5 micrometer; Pore Size: 100 angstrom;4.6mm x 15cm Length; Stainless Steel
Catalog No. 50-851-680