Learn More
Tosoh Bioscience LLC TSKGEL ODS-100V 4.6X250 5UM
Supplier: Tosoh Bioscience LLC 0021456
Column, Reversed Phase Chromatography; Tosoh TSKgel ODS-100V; Media: Silica; Particle: 5 micrometer; Pore Size: 100 angstrom;4.6mm x 25cm Length; Stainless Steel