Learn More
Tosoh Bioscience LLC TSKGEL ODS-100Z 2X150 5UM
Supplier:  Tosoh Bioscience LLC 0021459
Encompass
Column, Reversed Phase Chromatography; Tosoh TSKgel ODS-100Z; Media: Silica; Particle: 5 micrometer; Pore Size: 100 angstrom;2mm x 15cm Length; Stainless Steel
Catalog No. 50-851-684
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.