Learn More
Tosoh Bioscience LLC TSKGEL ODS-100Z 4.6X150 5UM
Catalog No. 50851686 Shop All Tosoh Bioscience LLC Products
Encompass
Column, Reversed Phase Chromatography; Tosoh TSKgel ODS-100Z; Media: Silica; Particle: 5 micrometer; Pore Size: 100 angstrom;4.6mm x 15cm Length; Stainless Steel
Catalog No. 50-851-686 Supplier Tosoh Bioscience LLC Supplier No. 0021461
Column, Reversed Phase Chromatography; Tosoh TSKgel ODS-100Z; Media: Silica; Particle: 5 micrometer; Pore Size: 100 angstrom;4.6mm x 15cm Length; Stainless Steel