Learn More
Tosoh Bioscience LLC TSKGEL ODS-120A 7.8X300
Supplier:  Tosoh Bioscience LLC 0007129
Encompass
Column, Reversed Phase Chromatography; Tosoh TSKgel ODS-120A; Media: Silica; Particle: 10 micrometer; Pore Size: 120 angstrom; 7.8mm I.D. x 30cm Length; Stainless Steel
Catalog No. 50-851-162