Learn More
Tosoh Bioscience LLC TSKGEL ODS-120T 21.5X300
Supplier: Tosoh Bioscience LLC 0007134
Column, Reversed Phase Chromatography; Tosoh TSKgel ODS-120T; Media: Silica; Particle: 10 micrometer; Pore Size: 120 angstrom; 21.5mm I.D. x 30cm Length; Stainless Steel