Learn More
Tosoh Bioscience LLC TSKGEL ODS-120T 4.6X150
Supplier:  Tosoh Bioscience LLC 0007637
Encompass
Column, Reversed Phase Chromatography; Tosoh TSKgel ODS-120T; Media: Silica; Particle: 5 micrometer; Pore Size: 120 angstrom; 4.6mm I.D. x 15cm Length; Stainless Steel
Catalog No. 50-851-216