Learn More
Tosoh Bioscience LLC TSKGEL ODS-120T GUARD 3PK
Catalog No. 50851526 Shop All Tosoh Bioscience LLC Products
Encompass
Cartridge, Guard; For 4.6mm I.D. Tosoh TSKgel ODS-120T columns; Media: Silica; 3.2mm I.D. x 1.5cm Length; 3/Pk
Catalog No. 50-851-526 Supplier Tosoh Bioscience LLC Supplier No. 0019006
Cartridge, Guard; For 4.6mm I.D. Tosoh TSKgel ODS-120T columns; Media: Silica; 3.2mm I.D. x 1.5cm Length; 3/Pk