Learn More
Tosoh Bioscience LLC TSKGEL ODS-80TM 21.5X300
Supplier: Tosoh Bioscience LLC 0014002
Column, Reversed Phase Chromatography; Tosoh TSKgel ODS-80Tm; Media: Silica; Particle: 10 micrometer; Pore Size: 80 angstrom; 21.5mm I.D. x 30cm Length; Stainless Steel