Learn More
Tosoh Bioscience LLC TSKGEL ODS-80TS 21.5X300
Catalog No. 50851451 Shop All Tosoh Bioscience LLC Products
Encompass
Column, Reversed Phase Chromatography; Tosoh TSKgel ODS-80Ts; Media: Silica; Particle: 10 micrometer; Pore Size: 80 angstrom; 21.5mm I.D. x 30cm Length; Stainless Steel - Standard
Catalog No. 50-851-451 Supplier Tosoh Bioscience LLC Supplier No. 0017380
Add to Cart
Add to Cart
Request a Quote
Column, Reversed Phase Chromatography; Tosoh TSKgel ODS-80Ts; Media: Silica; Particle: 10 micrometer; Pore Size: 80 angstrom; 21.5mm I.D. x 30cm Length; Stainless Steel - Standard