Learn More
Tosoh Bioscience LLC TSKGEL ODS-80TS 2X150
Supplier:  Tosoh Bioscience LLC 0018150
Encompass
Column, Reversed Phase Chromatography; Tosoh TSKgel ODS-80Ts; Media: Silica; Particle: 5 micrometer; Pore Size: 80 angstrom; 2mm I.D. x 15cm Length; Mini-Bore
Catalog No. 50-851-474