Learn More
Tosoh Bioscience LLC TSKGEL ODS-80TS GUARD 3PK
Supplier:  Tosoh Bioscience LLC 0019325
Encompass
Cartridge, Guard; For 2mm I.D. Tosoh TSKgel ODS columns; Media: Silica; 2mm I.D. x 1cm Length; 3/Pk
Catalog No. 50-851-556