Learn More
Tosoh Bioscience LLC TSKGEL ODS-80TS GUARD 3PK
Supplier:  Tosoh Bioscience LLC 0019011
Encompass
Cartridge, Guard; For 4.6mm I.D. Tosoh TSKgel ODS-80Ts columns; Media: Silica; 3.2mm I.D. x 1.5cm Length; 3/Pk
Catalog No. 50-851-530