Learn More
Tosoh Bioscience LLC TSKGEL ODS-80TS QA 4.6X75
Supplier:  Tosoh Bioscience LLC 0018519
Encompass
Column, Reversed Phase Chromatography; Tosoh TSKgel ODS-80Ts QA; Media: Silica; Particle: 5 micrometer; 4.6mm x 7.5cm Length; Quality Assurance
Catalog No. 50-851-505