Learn More
Tosoh Bioscience LLC TSKGEL OLIGODNA RP 7.8X150
Catalog No. 50550506 Shop All Tosoh Bioscience LLC Products
Encompass
LC C18 Column, TSKgel OligoDNA RP, 7.8 mm ID x 15 cm, 5 um
Catalog No. 50-550-506 Supplier Tosoh Bioscience LLC Supplier No. 0013353
LC C18 Column, TSKgel OligoDNA RP, 7.8 mm ID x 15 cm, 5 um