Learn More
Tosoh Bioscience LLC TSKGEL PHEN-5PW RP GUARD 3PK
Supplier:  Tosoh Bioscience LLC 0019007
Encompass
Cartridge, Guard; For 4.6mm I.D. Tosoh TSKgel Phenyl-5PW RP columns; Media: Silica; 3.2mm I.D. x 1.5cm Length; 3/Pk
Catalog No. 50-851-527