Learn More
Tosoh Bioscience LLC TSKGEL PHENYL-5PW (20) 250ML
Supplier:  Tosoh Bioscience LLC 0014718
Encompass
Media, Preparative Bulk Resins; For Hydrophobic Interaction Chromatography; Tosoh TSKgel Phenyl-5PW (20); Matrix: Polymer; Container Size: 250mL
Catalog No. 50-851-376