Learn More
Tosoh Bioscience LLC TSKGEL PHENYL-5PW (30) 250ML
Supplier:  Tosoh Bioscience LLC 0014720
Encompass
Media, Preparative Bulk Resins; For Hydrophobic Interaction Chromatography; Tosoh TSKgel Phenyl-5PW (30); Matrix: Polymer; Container Size: 250mL
Catalog No. 50-851-377