Learn More
Tosoh Bioscience LLC TSKGEL PHENYL-5PW 7.5X75
Supplier:  Tosoh Bioscience LLC 0007573
Encompass
Column, Hydrophobic Interaction Chromatography; Tosoh TSKgel Phenyl-5PW; Media: Polymer; Particle: 10 micrometer; Pore Size: 1000 angstrom; 7.5mm I.D. x 7.5cm Length; Stainless Steel
Catalog No. 50-851-211