Learn More
Tosoh Bioscience LLC TSKGEL PHENYL-5PW GL 8X75
Catalog No. 50851288 Shop All Tosoh Bioscience LLC Products
Encompass
Column, Hydrophobic interaction chromatography; Tosoh; TSKgel; Phenyl-5PW; Glass; Media: polymer; Particle: 10u m Pore Size: 1000angstrom pH stability: 2.0 to 12.0; 8mm I.D. x 7.5cm length
Catalog No. 50-851-288 Supplier Tosoh Bioscience LLC Supplier No. 0008804
Column, Hydrophobic interaction chromatography; Tosoh; TSKgel; Phenyl-5PW; Glass; Media: polymer; Particle: 10u m Pore Size: 1000angstrom pH stability: 2.0 to 12.0; 8mm I.D. x 7.5cm length