Learn More
Tosoh Bioscience LLC TSKGEL PHENYL-5PW RP 2X10 3PK
Supplier:  Tosoh Bioscience LLC 0042159
Encompass
Cartridge, Guard; For 2mm I.D. Tosoh TSKgel Phenyl-5PW RP column; Media: Polymer; 2mm I.D. x 1cm Length; 3/Pk
Catalog No. 50-851-707