Learn More
Tosoh Bioscience LLC TSKGEL PHENYL-5PW RP 2X75
Supplier:  Tosoh Bioscience LLC 0018756
Encompass
Column, Reversed Phase Chromatography; Tosoh TSKgel Phenyl-5PW RP; Media: Polymer; Particle: 10 micrometer; Pore Size: 1000 angstrom; 2mm I.D. x 7.5cm Length; Stainless Steel
Catalog No. 50-851-515