Learn More
Tosoh Bioscience LLC TSKGEL PHENYL-5PW RP 4.6X75
Catalog No. 50851261 Shop All Tosoh Bioscience LLC Products
Encompass
Column, Reversed Phase Chromatography; Tosoh TSKgel Phenyl-5PW RP; Media: Polymer; Particle: 10 micrometer; Pore Size: 1000 angstrom; 4.6mm I.D. x 7.5cm Length; Stainless Steel
Catalog No. 50-851-261 Supplier Tosoh Bioscience LLC Supplier No. 0008043
Column, Reversed Phase Chromatography; Tosoh TSKgel Phenyl-5PW RP; Media: Polymer; Particle: 10 micrometer; Pore Size: 1000 angstrom; 4.6mm I.D. x 7.5cm Length; Stainless Steel