Learn More
Tosoh Bioscience LLC TSKGEL PHENYL-5PW RP GL 8X75
Supplier:  Tosoh Bioscience LLC 0014007
Encompass
Column, Reversed phase chromatography; Tosoh; TSKgel Phenyl-5PW RP; Glass; Material: polymethacrylate; Particle: 10u m; Pore size: 1000angstrom pH: 2.0 to 12.0; 8mm I.D. x 7.5cm length
Catalog No. 50-851-317