Learn More
Tosoh Bioscience LLC TSKGEL QAE-2SW 4.6X250
Supplier:  Tosoh Bioscience LLC 0007166
Encompass
Column, Ion exchange chromatography; Tosoh; TSKgel QAE-2SW; Matrix: silica; Stainless-steel; Counter ion: H2; PO4 sup- Functional group: trimethylamino; Particle: 5u m; Pore size: 125
Catalog No. 50-851-173