Learn More
Tosoh Bioscience LLC TSKGEL SAX 6X150 5UM
Supplier:  Tosoh Bioscience LLC 0007157
Encompass
Column, Ion Exchange Chromatography; Anion Exchange; Tosoh TSKgel SAX, 5 micrometer; Media: Polymer; 6mm I.D. x 15cm Length; Stainless Steel
Catalog No. 50-851-166