Learn More
Tosoh Bioscience LLC TSKGEL SCX 7.8X300 5UM
Supplier:  Tosoh Bioscience LLC 0007158
Encompass
Column, Ion exchange chromatography; Tosoh; TSKgel SCX; Matrix: PS-DVB; Stainless-steel; Counter ion: Hsup+ Size: 5u m; (particle), 60angstrom (pore); Functional group: sulfonic acid; 7.8mm I.D. x 30cm length
Catalog No. 50-851-167