Learn More
Tosoh Bioscience LLC TSKGEL SP-2SW 4.6X250
Supplier:  Tosoh Bioscience LLC 0007165
Encompass
Column, Ion exchange chromatography; Tosoh; TSKgel SP-2SW; Matrix: spherical silica; Stainless-steel; Counter ion: Nasup+ Particle: 5u m; Pore size: 125angstrom pH: 2.0 -7.5; 4.6mm I.D. x 25cm length
Catalog No. 50-851-172