Learn More
Tosoh Bioscience LLC TSKGEL SP-5PW GL 8X75
Supplier:  Tosoh Bioscience LLC 0008803
Encompass
Column, Ion exchange chromatography; Tosoh; TSKgel SP-5PW; Matrix: hydroxylated methacrylic polymer; Glass; Counter ion: Nasup+ Particle: 10u m; Pore size: 1000angstrom pH: 2.0 -12; 8mm I.D. x 7.5cm length
Catalog No. 50-851-287