Learn More
Tosoh Bioscience LLC TSKGEL SUGAR AXG 4.6X150
Supplier:  Tosoh Bioscience LLC 0008640
Encompass
Column, Ion exchange chromatography; Tosoh; TSKgel sugar AXG; Matrix: PS-DVB polymer; Stainless-steel; Counter ion: HBO3 sup- Particle: 10u m; Pore size: 60angstrom Functional group: trimethyla
Catalog No. 50-851-275