Learn More
Tosoh Bioscience LLC TSKGEL SUGAR AXI 4.6X150
Supplier:  Tosoh Bioscience LLC 0008639
Encompass
Column, Ion exchange chromatography; Tosoh; TSKgel sugar AXI; Matrix: PS-DVB polymer; Stainless-steel; Counter ion: HBO3 sup- Particle: 8u m; Pore size: 60angstrom Functional group: trimethylam
Catalog No. 50-851-274