Learn More
Tosoh Bioscience LLC TSKGEL SUPER-ODS GUARD HOLDER
Supplier:  Tosoh Bioscience LLC 0018206
Encompass_Preferred
Holder, Guard Filter; For Tosoh TSKgel Super-ODS, -Octyl, and -Phenyl guard filters; 4mm I.D. x 0.4cm Length
Catalog No. 50-851-480