Learn More
Tosoh Bioscience LLC TSKGEL SUPERAW-H GRD 4.6X3.5
Supplier:  Tosoh Bioscience LLC 0019322
Encompass
Column, Guard; For Tosoh TSKgel SuperAW5000 - SuperAWM-H; Media: Polymer; 4.6mm I.D. x 3.5cm Length
Catalog No. 50-851-555