Learn More
Tosoh Bioscience LLC TSKGEL SUPERAW-L GUARD COLUMN
Supplier:  Tosoh Bioscience LLC 0019321
Encompass
Column, Guard; For Tosoh TSKgel SuperAW2500-4000; Media: Polymer; 4.6mm I.D. x 3.5cm Length
Catalog No. 50-851-554