Learn More
Tosoh Bioscience LLC TSKGEL SUPERAW3000 6X150
Supplier:  Tosoh Bioscience LLC 0019316
Encompass
Column, Size Exclusion Chromatography (SEC), Aqueous and Organic; Tosoh TSKgel SuperAW3000; Media: Polymer; Particle: 4 micrometer; 6mm I.D. x 15cm Length; Stainless Steel
Catalog No. 50-851-549